We offer a variety of universal-fit Invitrogen electroporation cuvettes for use with E. coli, yeast, and mammalian cells. Our electroporation cuvettes are available in several gap widths for optimal transformation efficiency for your specific cell type.
Advantages of Invitrogen electroporation cuvettes
Invitrogen electroporation cuvettes are precision built from high quality material to provide optimal transformation efficiency of foreign DNA into host bacterial, yeast, and mammalian cells. The cuvettes are compatible with all standard electroporation devices on the market. The advantages of these cuvettes include:
- The chamber design reduces the chance of introducing bubbles into the sample
- Allows direct recovery of cells in the cuvette
- Individually wrapped and gamma irradiated for sterility
- Color coded cuvette caps to identify gap widths
Electroporation cuvette selection guide
Invitrogen electroporation cuvettes are available in gap widths of 0.1 cm, 0.2 cm, and 0.4 cm; the cuvette caps are color coded for gap width. Table 1 lists the compatibility between the cuvette gap width and cell type. The cuvettes are sterilized with gamma irradiation, individually wrapped, and packaged in a bag of 50.
Table 1. Electroporation cuvette gap width and cell type suitability
0.1 cm cuvettes 25 kV/cm | 0.2 cm cuvettes 12.5 kV/cm | 0.4 cm cuvettes 6.25 kV/cm | |
---|---|---|---|
E. coli strains* | |||
MegaX DH10B T1R Electrocomp™ cells | – | ||
ElectroMAX DH10B T1R cells | – | ||
ElectroMAX DH10B cells | – | ||
TOP10 Electrocomp™ kit | – | ||
TOP10F’ Electrocomp™ kit | – | ||
ElectroMAX Stbl4 Competent Cells | – | ||
ElectroMAX DH12S Competent Cells | – | ||
ElectroMAX DH5α-E Competent Cells | – | ||
Yeast | – | – | |
Mammalian cells | – | – | |
Other eukaryotic cells | – | – | |
Order now | Order now | Order now |
Can you re-use electroporation cuvettes?
We do not recommend re-using electroporation cuvettes because washing cannot guarantee removal of all foreign DNA, and cuvettes cannot be easily sterilized. Also, metal plates are oxidized during the electroporation pulse so the same transformation efficiency cannot be achieved.
What are the minimal and maximal operating volumes?
- 1 mm gap cuvettes: 20 to 100 µL
- 2 mm gap cuvettes: 40 to 400 µL
- 4 mm gap cuvettes: 200 to 800 µL
Follow the optimized protocols for the cells and cuvettes that you are using.
Competent cells: Featured products
In addition to competent cells, we offer a broad suite of products for the cloning workflow.
Microbial
media
Explore types of bacterial growth media to use for cloning, plasmid DNA preparation, and protein expression.
Electroporation
cuvettes
Learn more about universal-fit electroporation cuvettes for use with competent cells.
TOPO PCR
cloning kits
Learn more about one-step PCR cloning that combines traditional restriction and ligation cloning.
Gibson Assembly
cloning kits
Gibson Assembly cloning kits provide highly efficient, seamless cloning, enabling the assembly of multiple DNA fragments of varying lengths into any vector.
Have any questions on competent cells or transformation? Click on the resources listed below to access overviews, videos, genotype guides, and educational resources.
Review rich educational content on transformation workflow, troubleshooting, strategies, and selection guides.
Stay up-to-date on industry trends in molecular cloning with on-demand webinars.
Short articles and digital handbook for cloning applications
Access white papers and technical and application notes on molecular cloning.
Watch instructional videos on troubleshooting tips, how-to videos, and explainers related to molecular cloning.
Get support for your cloning experiments