Targeted sequencing with Oncomine NGS oncology assays
Oncomine™ assays are multi-biomarker targeted assays designed for cancer research. Enabling next-generation sequencing analysis of multiple biomarker types—fusions, insertion/deletions (indels), single nucleotide variants, and copy number variations—Oncomine assays are part of an end-to-end workflow that includes simple, scalable sequencing with Ion S5™ Series sequencers, and optimized bioinformatics and reporting with the Oncomine™ Knowledgebase Reporter.
Whether you are working with DNA or RNA, formalin-fixed, paraffin-embedded (FFPE) tissue samples, or blood, Oncomine assays engender trust among clinical cancer researchers because:
- Content includes cancer genes and is reviewed by experienced professionals to help ensure the highest relevancy
- Protocols and verification are based on analyses of clinical samples to help ensure repeatable results
- Assays are produced under enhanced manufacturing quality control to facilitate consistent analyses across large sample sets
- Specialized application support teams are available to help enable faster implementation in your lab
Oncomine assays
Designed for concurrent DNA and RNA NGS analysis from FFPE samples. Targets 52 genes relevant to solid tumors. Detects SNVs, indels, CNVs, and fusions.
Designed for concurrent DNA and RNA NGS analysis from FFPE samples. Targets 143 cancer genes relevant to clinical cancer research. Detects SNVs, indels, CNVs, and fusions.
Tumor type-specific assays designed for NGS analysis from cell-free DNA. Targets genes relevant to lung, colon,* and breast* tumor samples. Detects SNVs and indels.
Powerful tools enabling complete NGS data analysis. Go from hundreds down to a few relevant cancer drivers in a report that enables contextual investigation of sample-specific variants.
With Ion Torrent™ next-generation sequencing (NGS) systems, you can analyze your sample for hundreds of variants of interest using a single sample in a single experiment. You can detect hotspots, SNVs, indels, CNVs, and gene fusions in a single run. For FFPE samples and liquid biopsy samples, both Ion AmpliSeq™ technology and Tag Sequencing technology enable results from as little as 1 ng of sample input.
With Ion Torrent NGS systems and Oncomine assays, your lab has the flexibility to scale and automate analyses to meet your sampling needs. Coupled with the Ion Chef™ System and the Ion S5 System, the Oncomine™ Focus Assay enables sample to refined variant data in two days. And this same easy-to-use workflow achieves automated speed and simplicity to enable your lab technicians to run samples consistently.
The Oncomine NGS oncology workflow enables you to go from hundreds of variants down to a few relevant cancer drivers. The comprehensive workflow includes powerful tools for cancer research, including a curated, verified knowledgebase that informs assay design, variant annotations to enhance your NGS data analysis, and a report that enables contextual investigation of sample-specific variants.
Resources
* The content provided herein may relate to products that have not been officially released and is subject to change without notice.