Traceable™ products
We offer an unsurpassed range of thermometers, timers, and stopwatches to meet the requirements of any laboratory. Each Traceable product is individually serialised, calibrated, and certified. An individually serialised Traceable Calibration Certificate provides you with the assurance that an independent auditor has checked the methods, procedures, testing, technique, and record-keeping practices of the calibration testing laboratory.
We have a wide range of affordable digital thermometers and innovative technology, such as bottle probe thermometers for refrigerators and freezers and infrared thermometers that suit any application.
Traceable™ timers and stopwatches are factory calibrated to NIST standards. With modern design and a wide variety from which to choose, Traceable timers and stopwatches have your every need covered.
Fisher Scientific TraceableLIVE™ Wi-Fi datalogging refrigerator/freezer thermometers with remote notification
Live and secure remote monitoring wherever you go
Simple to set up, TraceableLIVE products connect to your existing Wi-Fi network and transmit critical data to your personal cloud. The device can then be monitored through your smartphone, tablet, or computer.
Wirelessly monitor temperature, humidity, CO2 and barometric pressure.
Instant notification of alerts through mobile push, email or text.
Event documentation to track alarm events, low battery or loss of Wi-Fi connectivity.
Documented change control for permanent records of events.
Unlimited scalability – monitor individual labs or an entire network of facilities.
Continuous data transmission & monitoring using SSL encrypted secure connection.