Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • BCL-W Antibodies

NeoBiotechnologies

BCL2-like 2 (BCL2L2)/BCL-W Monoclonal Antibody (PCRP-BCL2L2-1A4)

View all (18) BCL-W antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite BCL2-like 2 (BCL2L2)/BCL-W Monoclonal Antibody (PCRP-BCL2L2-1A4)

  • Antibody Testing Data (5)
BCL2-like 2 (BCL2L2)/BCL-W Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
BCL2-like 2 (BCL2L2)/BCL-W Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

BCL2-like 2 (BCL2L2)/BCL-W Antibody (599-MSM2-P1ABX) in ICC/IF

Immunofluorescence analysis of PFA-fixed MCF-7 cells stained using BCL2L2 Mouse Monoclonal Antibody (PCRP-BCL2L2-1A4) followed by goat anti-mouse IgG-CF488 (green). CF640R phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

BCL2-like 2 (BCL2L2)/BCL-W Antibody in Immunocytochemistry (ICC/IF)
BCL2-like 2 (BCL2L2)/BCL-W Antibody in Immunocytochemistry (ICC/IF)
BCL2-like 2 (BCL2L2)/BCL-W Antibody in Flow Cytometry (Flow)
BCL2-like 2 (BCL2L2)/BCL-W Antibody in Peptide array (ARRAY)
BCL2-like 2 (BCL2L2)/BCL-W Antibody in SDS-PAGE (SDS-PAGE)
BCL2-like 2 (BCL2L2)/BCL-W Monoclonal Antibody (PCRP-BCL2L2-1A4)

Product Details

599-MSM2-P1ABX

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

1-2 µg/mL
-

Flow Cytometry (Flow)

1-2 µg/10^6 cells
-

Immunoprecipitation (IP)

1-2 µg
-

Peptide Array (Array)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

PCRP-BCL2L2-1A4

Immunogen

Recombinant human full-length BCL2L2 protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Target Information

Using a PCR-based strategy, an additional protein with life-promoting activity, designated Bcl-w, has been identified. The protein is highly conserved between mouse and human and is encoded by a gene located near the TCRalpha gene on chromosome 14. Bcl-w is expressed in myeloid cell lines but not in T and B lymphocytes, and can be found in a wide range of tissues. An alternative splicing event in exon 4 results in two transcripts. The first, Bcl-w, encodes a protein of 193 amino acids, and the second, Bcl-w/rox, encodes a protein 333 amino acids in length. The "rox" portion of Bcl-w/rox shows a striking 66% amino acid sequence identity with the Drosophila rox2 protein; however, the Bcl-w/rox transcript may be expressed at very low levels.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Apoptosis regulator Bcl-W; Bcl-2-like protein 2; Bcl2-L-2; BCL2-like 2 protein; Bcl2-like protein 2; protein phosphatase 1, regulatory subunit 51

View more View less

Gene Aliases: BCL-W; BCL2-L-2; BCL2L2; BCLW; KIAA0271; PPP1R51

View more View less

UniProt ID: (Human) Q92843

View more View less

Entrez Gene ID: (Human) 599

View more View less

Function(s)
protein binding protein homodimerization activity protein heterodimerization activity BH domain binding
Process(es)
spermatogenesis intrinsic apoptotic signaling pathway in response to DNA damage negative regulation of apoptotic process Sertoli cell proliferation extrinsic apoptotic signaling pathway in absence of ligand negative regulation of intrinsic apoptotic signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0