">
Cell Proliferation Assays for Flow Cytometry
Find flow cytometry dyes, kits, and antibodies for cell proliferation
Using flow cytometry proliferation measurements are typically made based on average DNA content or on cellular metabolism parameters. Assays can report either total or live cell numbers or measure DNA synthesis in single cells. We offer dyes, kits, and antibodies to track proliferation. Use the selection guide below to find a reagent for flow cytometry assays or multi-color panels.
Cell proliferation assays for flow cytometry selection guide
Click-iT Edu assays for flow cytometry
Measure DNA synthesis with Click-iT flow cytometry kits. The kits are optimized for live cells, the click detection step comes after the fixation.
Product
Fluorescent label
Laser (nm)
Ex/Em (nm)
Multiplex options
Click-iT Plus Edu Alexa Fluor 350 Flow Cytometry Kit
Alexa Fluor 350 picolyl azide
UV
350/438
 Can be co-stained with standard organic dyes (FITC, Alexa Fluor dyes, etc.), R-PE, R-PE tandems, fluorescent proteins.
Click-iT Plus Edu Pacific Blue Flow Cytometry Kit
Pacific Blue picolyl azide
405
410/455
Click-iT Plus Edu Alexa Fluor 488 Flow Cytometry Kit
Alexa Fluor 488 picolyl azide
488
495/519
Click-iT Plus Edu Alexa Fluor 594 Flow Cytometry Kit
Alexa Fluor 594 picolyl azide
532 or 561
532 or 561/615
Click-iT Plus Edu Alexa Fluor 647 Flow Cytometry Kit
Alexa Fluor 647 picolyl azide
633/635
650/668
Click-iT Edu Pacific Blue Flow Cytometry Kit
Pacific Blue azide
405
410/455
Limited; can be co-stained with standard organic dyes.
Not compatible with R-PE, R-PE tandems, fluorescent proteins.
Click-iT Edu Alexa Fluor 488 Flow Cytometry Kit
Alexa Fluor 488 azide
488
495/519
Click-iT Edu Alexa Fluor 647 Flow Cytometry Kit
Alexa Fluor 647 azide
633/635
350/668
CellTrace proliferation kits for flow cytometry
CellTrace cell proliferation kits permanently label cells with fluorescent stains to trace generations or divisions in vivo or in vitro without affecting morphology or physiology. 
Click-iT Edu cell proliferation assays
A common method to measure DNA synthesis involves the incorporation of BrdU into newly synthesized DNA in living cells, which can later be detected using a fluorescently labeled antibody that recognizes BrdU. The fluorescent signal can be analyzed with flow cytometry instruments to provide a quantitative measurement of DNA synthesis during cell proliferation. 
Click-iT EdU flow cytometry assay kits were developed to overcome some of limitations of the BrdU technique. Click-iT EdU and Plus EdU flow cytometry assays utilize the power of click chemistry and the modified nucleoside EdU to offer an excellent alternative to BrdU staining for detecting and quantitating newly synthesized DNA.
  • Accuracy—typically better than BrdU assays with minimal variation (low CVs)
  • Simplicity—streamlined protocol
  • Compatibility—assays allow users to multiplex with GFP, mCherry, APC, PerCP, PE, and other fluorophores (Click-iT Plus EdU kits only)
Explore: Click-iT EdU cell proliferation assays
CellTrace cell proliferation kits
CellTrace stains are fluorescent tracking dyes that are retained in living cells. With each generation, the amount of dye in each cell is halved, so fluorescence analysis of proliferating populations can be used to quantify the successively decreasing brightness for each generation. These dyes are typically effective at reporting proliferation above background autofluorescence for over six generations.
  • Excellent performance—bright, single-peak staining enables visualization of multiple generations
  • Long-term signal stability—well retained in cells for several days post-stain
  • Non-cytotoxic—no known effect on proliferative ability or biology of cells
  • Compatibility—bright, single-peak staining available in multiple fluorophores
  • Versatile—multiple colors available to easily combine with antibodies or markers of cell function, such as GFP
Analysis of the level of fluorescence in the cell populations by flow cytometry permits the determination of the number of generations through which a cell has progressed since the label was applied (Figure 1).
Figure 1. Mechanism of cell labeling with CellTrace cell proliferation kits. (A) Illustration of proliferation analysis by dye dilution. (B) Flow cytometric analysis reveals a bright, homogenous fluorescent signal from the initial population of cells. Subsequent cell divisions result in larger numbers of cells, each with half the fluorescence intensity of its parent cell.
The CellTrace Cell Proliferation Kits are available for the UV, violet, blue, yellow (561 nm), and red lasers allowing researchers to analyze cell proliferation in combination with other live-cell applications (i.e., immunophenotyping, cell sorting, and cell-cycle analysis), maximizing the information that can be collected in a single experiment, with minimal compensation.
Antibodies for cell proliferation markers
Cell cycle phases coordinate multiple proteins to synthesize DNA and divide cells. Building flow cytometry panels for the detection of cell proliferation markers can help evaluate this process. Common cell proliferation markers include 5-bromodeoxyuridine (BrdU), Ki-67, minichromosome maintenance (MCM2), and proliferating cell nuclear antigen (PCNA).
Flow Cytometry Panel Design Service and Panel Builder Tool
Design flow cytometry panels free with scientists or using our easy tool.
Flow Cytometry Reagents
Explore viability dyes, Live/Dead assays for cell health and function, RNA detection by flow cytometry, and much more.
Antibodies for Flow Cytometry
Teaser: Explore more than 80 fluorophore dyes and 28,000+ conjugated antibodies.
Flow Cytometry Instruments
Explore our portfolio of Attune Flow Cytometers for flow cytometry analysis and the Bigfoot Spectral Cell Sorter for high-speed cell sorting.
Spectral Flow Cytometry Assays and Reagents
Explore next-level spectral flow cytometry technologies.
Cy™ is a trademark or registered trademark of GE Healthcare. 
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. 
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and is used under license. 
For Research Use Only. Not for use in diagnostic procedures. 
">