">
Invitrogen eFluor organic dyes are specifically designed for labeling and detecting biomolecules, providing exceptional optical performance and full compatibility with high-parameter flow cytometry. Featuring enhanced brightness and photostability, eFluor tandem fluorophores are replacements for traditional Cyanine dye–based tandem options. Named after their emission wavelength, eFluor dyes facilitate easy selection for your research needs. 
eFluor dye selection guide
Violet
(Ex/Em)
Green
(Ex/Em)
Yellow
(Ex/Em)
Red
(Ex/Em)
Near IR/IR
(Ex/EM)
eFluor 450 dye
eFluor 450 antibodies 
(405/445 nm)
eFluor 570 antibodies
(556/569 nm)
PE-eFluor 610 dye
PE-eFluor 610 antibodies 
(565/606 nm)
eFluor 660 dye
eFluor 660 antibodies 
(651/668 nm)
APC-eFluor 780 dye
APC-eFluor 780 antibodies 
(756/785 nm)
eFluor 506 dye
eFluor 506 antibodies 
(419/508 nm)
PerCP-eFluor 710 dye 
PerCP-eFluor 710 antibodies 
(482/708 nm)
eFluor 615 antibodies 
(595/614 nm)
Flow Cytometry Panel Design Service and Panel Builder Tool
Design flow cytometry panels free with scientists or using our easy tool.
Flow Cytometry Panel Design
Flow Cytometry Reagents
Explore viability dyes, Live/Dead assays for cell health and function, RNA detection by flow cytometry, and much more.
Beads for Compensation, Instrument Controls, and Standards
Explore flow cytometry beads for compensation, calibrations, size reference, and cell counting. 
Flow Cytometry Instruments
Explore our portfolio of Attune Flow Cytometers for flow cytometry analysis and the Bigfoot Spectral Cell Sorter for high-speed cell sorting.
Spectral Flow Cytometry Assays and Reagents
Explore next-level spectral flow cytometry technologies.
Cy™ is a trademark or registered trademark of GE Healthcare. 
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. 
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and is used under license. 
For Research Use Only. Not for use in diagnostic procedures. 
">