Assay Details
Gene Symbol: LOC100284883 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157778.1 | NP_001151250.1 | 448 | 110 | ||
GenBank mRNA | EU958920.1 | - | 365 | 110 | ||
EU970014.1 | - | 448 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
