This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Zm04034001_gH
See other LOC100282425 GE Assays ›
Gene Symbol:
LOC100282425
Gene Name:
protein binding protein
Gene Aliases:
ZEAMMB73_327370
Chromosome Location:
-
Species:
Zea mays
Species Specific ID (Flybase ID):
-
Assay ID Zm04034001_gH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100282425
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001155336.1 NP_001148808.1 579 87
GenBank mRNA BT068780.1 - 501 87
EU960851.1 - 579 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Zea mays

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0