This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03723778_m1
See other AGPAT1 GE Assays ›
Gene Symbol:
agpat1
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases:
1-agpat1, g15, lpaat-alpha, lpaata
Chromosome Location:
Chr.8: 54298648 - 54329404 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03723778_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: agpat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001097333.1 NP_001090802.1 7 - 8 930 69
XM_012967818.2 XP_012823272.1 8 - 9 919 69
XM_012967820.2 XP_012823274.1 7 - 8 847 69
XM_012967821.2 XP_012823275.1 7 - 8 795 69
XM_012967822.2 XP_012823276.1 8 - 9 861 69
XM_012967823.2 XP_012823277.1 8 - 9 920 69
XM_012967824.2 XP_012823278.1 8 - 9 809 69
XM_012967825.2 XP_012823279.1 7 - 8 776 69
XM_012967826.2 XP_012823280.1 6 - 7 723 69
XM_012967827.2 XP_012823281.1 7 - 8 837 69
XM_018096082.1 XP_017951571.1 8 - 9 809 69
XM_018096083.1 XP_017951572.1 6 - 7 908 69
XM_018096084.1 XP_017951573.1 6 - 7 699 69
GenBank mRNA BC135226.1 - 7 - 8 930 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0