This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03721507_m1
See other SLC25A39 GE Assays ›
Gene Symbol:
slc25a39
Gene Name:
solute carrier family 25 member 39
Gene Aliases:
-
Chromosome Location:
Chr.10: 9189803 - 9203655 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03721507_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc25a39
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083349.1 NP_001076818.1 9 - 10 815 82
XM_012971713.2 XP_012827167.1 8 - 9 1256 82
XM_012971714.2 XP_012827168.1 8 - 9 852 82
XM_012971715.2 XP_012827169.1 8 - 9 802 82
GenBank mRNA BC088076.1 - 7 - 8 707 82
BC135927.1 - 9 - 10 815 82
BC155021.1 - 9 - 10 866 82
BC161586.1 - 9 - 10 866 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0