This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03719359_g1
See other AURKB GE Assays ›
Gene Symbol:
aurkb
Gene Name:
aurora kinase B
Gene Aliases:
AurB, Aurora-B, XAIRK2, aik2, aim-1, aim1, airk2, ark2, aurkb-a, aurkb-b, ipl1, stk12, stk5
Chromosome Location:
Chr.3: 131759450 - 131768598 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03719359_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: aurkb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016859.3 NP_001016859.2 3 - 4 236 91
XM_012953827.2 XP_012809281.1 2 - 3 247 91
XM_012953828.2 XP_012809282.1 3 - 4 282 91
GenBank mRNA BC135173.1 - 3 - 4 236 91
CR848658.2 - 3 - 4 156 91
CT027921.1 - 2 - 3 197 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0