Assay Details
Gene Symbol: thul16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011392.1 | NP_001011392.1 | 5 - 6 | 836 | 74 | |
XM_018096243.1 | XP_017951732.1 | 5 - 6 | 361 | 74 | ||
XM_018096244.1 | XP_017951733.1 | 5 - 6 | 603 | 74 | ||
GenBank mRNA | BC088584.1 | - | 5 - 6 | 836 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
