This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03685286_m1
See other HNRNPC GE Assays ›
Gene Symbol:
hnrnpc
Gene Name:
heterogeneous nuclear ribonucleoprotein C (C1/C2)
Gene Aliases:
hnrnp, hnrpc, snrpc
Chromosome Location:
Chr.1: 131091582 - 131105599 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03685286_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hnrnpc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005088.2 NP_001005088.1 3 - 4 439 63
XM_012960151.2 XP_012815605.1 3 - 4 1024 63
XM_012960155.2 XP_012815609.1 3 - 4 1152 63
XM_012960157.2 XP_012815611.1 2 - 3 472 63
XM_018092371.1 XP_017947860.1 3 - 4 1025 63
GenBank mRNA BC077013.1 - 3 - 4 440 63
CT027976.2 - 3 - 4 373 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0