This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Vv04086037_m1
See other AP1 GE Assays ›
Gene Symbol:
AP1
Gene Name:
apetala1
Gene Aliases:
GSVIVT00030005001
Chromosome Location:
Chr.1: 134931 - 160101 on Build 12X
Species:
Vitis vinifera
Species Specific ID (Flybase ID):
-
Assay ID Vv04086037_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AY538746.1 - 5 - 6 643 89
GU133634.1 - 5 - 6 477 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Vitis vinifera Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0