This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06942779_g1
See other RSRP1 GE Assays ›
Gene Symbol:
RSRP1
Gene Name:
arginine and serine rich protein 1
Gene Aliases:
-
Chromosome Location:
Chr.6: 82850947 - 82861578 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06942779_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RSRP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243642.1 NP_001230571.1 4 - 5 958 103
XM_021093409.1 XP_020949068.1 1213 103
XM_021093410.1 XP_020949069.1 1283 103
XM_021093411.1 XP_020949070.1 1243 103
XM_021093412.1 XP_020949071.1 1307 103
XM_021093413.1 XP_020949072.1 1153 103
XM_021093414.1 XP_020949073.1 1247 103
XM_021093415.1 XP_020949074.1 1064 103
GenBank mRNA AK233814.1 - 4 - 5 958 103
AK234227.1 - 4 - 5 949 103
AK234378.1 - 4 - 5 946 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0