This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06942380_s1
See other RNASE1 GE Assays ›
Gene Symbol:
RNASE1
Gene Name:
ribonuclease A family member 1, pancreatic
Gene Aliases:
RNS1
Chromosome Location:
Chr.7: 78199007 - 78214832 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06942380_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RNASE1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001167655.2 NP_001161127.1 1 425 63
XM_021098159.1 XP_020953818.1 783 63
XM_021098160.1 XP_020953819.1 785 63
XM_021098161.1 XP_020953820.1 470 63
GenBank mRNA AK240286.1 - 2 489 63
AK399861.1 - 2 464 63
AY610181.1 - 2 460 63
EU814500.1 - 1 428 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

RNA phosphodiester bond hydrolysis, endonucleolytic
nucleic acid binding
ribonuclease A activity
lyase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0