This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06941640_m1
See other LIME1 GE Assays ›
Gene Symbol:
LIME1
Gene Name:
Lck interacting transmembrane adaptor 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 62652618 - 62655342 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06941640_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIME1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244490.1 NP_001231419.1 3 - 4 407 95
XM_021077271.1 XP_020932930.1 1568 95
XM_021077272.1 XP_020932931.1 1499 95
XM_021077273.1 XP_020932932.1 753 95
GenBank mRNA AK236913.1 - 3 - 4 407 95
AK345604.1 - 3 - 4 421 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0