This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06940523_s1
See other ILDR2 GE Assays ›
Gene Symbol:
ILDR2
Gene Name:
immunoglobulin like domain containing receptor 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 84056736 - 84117123 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06940523_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ILDR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021089477.1 XP_020945136.1 496 97
XM_021089478.1 XP_020945137.1 299 97
XM_021089479.1 XP_020945138.1 285 97
XM_021089480.1 XP_020945139.1 497 97
XM_021089482.1 XP_020945141.1 402 97
XM_021089483.1 XP_020945142.1 516 97
XM_021089484.1 XP_020945143.1 487 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0