This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06926832_m1
See other GPR132 GE Assays ›
Gene Symbol:
GPR132
Gene Name:
G protein-coupled receptor 132
Gene Aliases:
-
Chromosome Location:
Chr.Unknown: 3212828 - 3225182 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06926832_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR132
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005656481.3 XP_005656538.1 485 70
XM_021081565.1 XP_020937224.1 488 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005656481.3, XM_021081565.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Sus scrofa

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0