This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06920485_m1
See other APOH GE Assays ›
Gene Symbol:
APOH
Gene Name:
apolipoprotein H
Gene Aliases:
-
Chromosome Location:
Chr.12: 12827195 - 12839172 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06920485_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003131266.4 XP_003131314.1 122 101
XM_005668662.3 XP_005668719.1 122 101
GenBank mRNA AK232456.1 - 1 - 2 99 101
AK232894.1 - 1 - 2 94 101
AK233615.1 - 1 - 2 97 101
AK346407.1 - 1 - 2 97 101
AK393969.1 - 1 - 2 125 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0