This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06919964_m1
See other RNF2 GE Assays ›
Gene Symbol:
RNF2
Gene Name:
ring finger protein 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 126172637 - 126284567 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06919964_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RNF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003130379.6 XP_003130427.2 804 95
XM_005667821.3 XP_005667878.2 506 95
XM_005667825.3 XP_005667882.2 486 95
XM_013979932.2 XP_013835386.2 878 95
XM_021102630.1 XP_020958289.1 486 95
XM_021102631.1 XP_020958290.1 900 95
XM_021102632.1 XP_020958291.1 600 95
XM_021102633.1 XP_020958292.1 487 95
XM_021102634.1 XP_020958293.1 490 95
XM_021102635.1 XP_020958294.1 486 95
GenBank mRNA AK232204.1 - 5 - 6 880 95
AK239504.1 - 4 - 5 562 95
AK397479.1 - 4 - 5 742 95
AY610011.1 - 4 - 5 526 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0