This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06916660_m1
See other SGCG GE Assays ›
Gene Symbol:
SGCG
Gene Name:
sarcoglycan gamma
Gene Aliases:
-
Chromosome Location:
Chr.11: 2310280 - 2380054 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06916660_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SGCG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013980394.2 XP_013835848.2 475 54
XM_021065454.1 XP_020921113.1 595 54
XM_021065455.1 XP_020921114.1 595 54
XM_021065456.1 XP_020921115.1 695 54
XM_021065457.1 XP_020921116.1 695 54
XM_021065458.1 XP_020921117.1 505 54
XM_021065459.1 XP_020921118.1 584 54
XM_021065460.1 XP_020921119.1 684 54
XM_021065461.1 XP_020921120.1 586 54
GenBank mRNA FJ481171.1 - 4 - 5 411 54

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header