This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06915608_m1
See other INTS6L GE Assays ›
Gene Symbol:
INTS6L
Gene Name:
integrator complex subunit 6 like
Gene Aliases:
-
Chromosome Location:
Chr.X: 111011860 - 111072939 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06915608_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: INTS6L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003135419.4 XP_003135467.1 2026 73
XM_013991006.2 XP_013846460.1 1759 73
XM_013991007.2 XP_013846461.1 2138 73
XM_013991008.2 XP_013846462.1 1935 73
XM_013991009.2 XP_013846463.1 1648 73
XM_013991010.2 XP_013846464.1 2277 73
XM_013991011.2 XP_013846465.1 1609 73
XM_013991012.2 XP_013846466.1 1988 73
XM_013991013.2 XP_013846467.1 1658 73
XM_021080861.1 XP_020936520.1 2142 73
XM_021080863.1 XP_020936522.1 1822 73
XM_021080864.1 XP_020936523.1 1933 73
XM_021080866.1 XP_020936525.1 1648 73
XM_021080867.1 XP_020936526.1 2025 73
XM_021080868.1 XP_020936527.1 1876 73
XM_021080869.1 XP_020936528.1 1783 73
GenBank mRNA AK398507.1 - 4 - 5 526 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

RNA helicase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0