This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06907453_s1
See other ARHGAP5 GE Assays ›
Gene Symbol:
ARHGAP5
Gene Name:
Rho GTPase activating protein 5
Gene Aliases:
-
Chromosome Location:
Chr.7: 67494237 - 67631461 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06907453_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013978128.2 XP_013833582.2 6463 131
XM_021099176.1 XP_020954835.1 6457 131
XM_021099177.1 XP_020954836.1 6274 131
XM_021099178.1 XP_020954837.1 6275 131
XM_021099179.1 XP_020954838.1 6274 131
XM_021099180.1 XP_020954839.1 6414 131
XM_021099182.1 XP_020954841.1 6461 131
GenBank mRNA AK391938.1 - 1 118 131

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0