This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06900965_s1
See other LYN GE Assays ›
Gene Symbol:
LYN
Gene Name:
LYN proto-oncogene, Src family tyrosine kinase
Gene Aliases:
-
Chromosome Location:
Chr.4: 75793273 - 75903710 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06900965_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LYN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021089356.1 XP_020945015.1 4626 89
XM_021089357.1 XP_020945016.1 4627 89
XM_021089358.1 XP_020945017.1 4696 89
XM_021089359.1 XP_020945018.1 4823 89
XM_021089360.1 XP_020945019.1 5047 89
XM_021089361.1 XP_020945020.1 4627 89
XM_021089362.1 XP_020945021.1 4626 89
XM_021089363.1 XP_020945022.1 4816 89
GenBank mRNA AK350933.1 - 1 1218 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm