Assay Details
Gene Symbol: FAM71D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK344096.1 | - | 1 | 1717 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Gene Symbol: FAM71D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK344096.1 | - | 1 | 1717 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |