This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04954895_m1
See other CKMT1A GE Assays ›
Gene Symbol:
CKMT1A
Gene Name:
creatine kinase, mitochondrial 1A
Gene Aliases:
CKMT1, CKMT1B, Mia-CK, U-MtCK
Chromosome Location:
Chr.1: 127852533 - 127859066 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04954895_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CKMT1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001315638.1 NP_001302567.1 4 - 5 666 88
XM_003121548.3 XP_003121596.1 952 88
XM_005659675.3 XP_005659732.1 989 88
XM_005659676.3 XP_005659733.1 982 88
XM_013993080.1 XP_013848534.1 913 88
XM_021074896.1 XP_020930555.1 971 88
XM_021074950.1 XP_020930609.1 965 88
XM_021074988.1 XP_020930647.1 1067 88
XM_021075051.1 XP_020930710.1 967 88
GenBank mRNA AK231576.1 - 5 - 6 941 88
AK345070.1 - 5 - 6 855 88
AK350124.1 - 5 - 6 952 88
AK396391.1 - 5 - 6 945 88
AK399926.1 - 4 - 5 863 88
EU650786.1 - 4 - 5 666 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

amino acid kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphorylation
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0