This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04652610_s1
See other MIR9825 GE Assays ›
Gene Symbol:
MIR9825
Gene Name:
microRNA mir-9825
Gene Aliases:
ssc-mir-9825
Chromosome Location:
Chr.4: 84110767 - 84110829 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04652610_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR9825
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_128510.1 1 28 57

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream immunoglobulin like domain containing receptor 2 ILDR2 Sense Chr. 4 - 84056736 - 84117123 on NCBI Build Sscrofa11.1
Downstream transcriptional adaptor 1 TADA1 63.7kb Sense Chr. 4 - 84174481 - 84190077 on NCBI Build Sscrofa11.1

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0