Assay Details
Gene Symbol: PRXL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244373.2 | NP_001231302.1 | 6 - 7 | 895 | 108 | |
NM_001244374.2 | NP_001231303.1 | 5 - 6 | 781 | 108 | ||
NM_001244375.2 | NP_001231304.1 | 5 - 6 | 764 | 108 | ||
NM_001359936.1 | NP_001346865.1 | 6 - 7 | 878 | 108 | ||
NM_001359937.1 | NP_001346866.1 | 5 - 6 | 614 | 108 | ||
NM_001359938.1 | NP_001346867.1 | 5 - 6 | 631 | 108 | ||
NM_001359939.1 | NP_001346868.1 | 6 - 7 | 761 | 108 | ||
XM_005671141.3 | XP_005671198.1 | 692 | 108 | |||
GenBank mRNA | AK232037.1 | - | 6 - 7 | 812 | 108 | |
AK233264.1 | - | 5 - 6 | 685 | 108 | ||
AK234200.1 | - | 5 - 6 | 699 | 108 | ||
AK389174.1 | - | 5 - 6 | 724 | 108 | ||
AK390968.1 | - | 5 - 6 | 754 | 108 | ||
AK400395.1 | - | 6 - 7 | 878 | 108 | ||
AY610040.1 | - | 4 - 5 | 832 | 108 | ||
DY420282.1 | - | 1 - 2 | 89 | 108 | ||
EV914221.2 | - | 1 - 2 | 115 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
