This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04325813_m1
See other CD79B GE Assays ›
Gene Symbol:
CD79B
Gene Name:
CD79b molecule
Gene Aliases:
-
Chromosome Location:
Chr.12: 15052966 - 15056441 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04325813_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD79B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243912.1 NP_001230841.1 1 - 2 126 100
XM_021066088.1 XP_020921747.1 167 100
GenBank mRNA AK237283.1 - 1 - 2 136 100
AK397726.1 - 1 - 2 145 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0