Assay Details
Gene Symbol: ESRRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170521.1 | NP_001163992.1 | 3 - 4 | 573 | 91 | |
XM_005660695.3 | XP_005660752.2 | 794 | 91 | |||
XM_021077409.1 | XP_020933068.1 | 604 | 91 | |||
GenBank mRNA | FJ446485.1 | - | 3 - 4 | 573 | 91 | |
KP717445.1 | - | 3 - 4 | 573 | 91 | ||
KP717446.1 | - | 4 - 5 | 506 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
