This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03389943_g1
See other SLA-DRA GE Assays ›
Gene Symbol:
SLA-DRA
Gene Name:
MHC class II DR-alpha
Gene Aliases:
SLA-DR-alpha, SLA-DRA1, SLA-DRA2
Chromosome Location:
Chr.7: 24825037 - 24830650 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03389943_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLA-DRA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113706.1 NP_001107178.1 2 - 3 397 64
GenBank mRNA AB215118.1 - 2 - 3 320 64
AK239665.1 - 2 - 3 410 64
AK351896.1 - 2 - 3 401 64
AK389739.1 - 2 - 3 420 64
AK397711.1 - 2 - 3 398 64
AK398000.1 - 2 - 3 402 64
AK398178.1 - 2 - 3 396 64
AK398242.1 - 2 - 3 399 64
AK398994.1 - 2 - 3 399 64
AY243102.1 - 2 - 3 320 64
AY243106.1 - 2 - 3 320 64
AY247781.1 - 2 - 3 342 64
AY247782.1 - 2 - 3 342 64
AY285926.1 - 2 - 3 322 64
AY285928.1 - 2 - 3 320 64
AY285929.1 - 2 - 3 320 64
AY285933.1 - 2 - 3 322 64
AY285935.1 - 2 - 3 322 64
AY285937.1 - 2 - 3 320 64
AY285938.2 - 2 - 3 320 64
AY459302.2 - 2 - 3 320 64
DQ298395.1 - 2 - 3 320 64
DQ883222.1 - 2 - 3 342 64
DQ883223.1 - 2 - 3 342 64
DQ883224.1 - 2 - 3 342 64
EF143987.1 - 2 - 3 336 64
EU432070.1 - 2 - 3 322 64
EU432071.1 - 2 - 3 322 64
EU722916.1 - 2 - 3 346 64
EU722917.1 - 2 - 3 346 64
EU722918.1 - 2 - 3 343 64
EU728595.1 - 2 - 3 336 64
FJ905823.1 - 2 - 3 371 64
FJ905824.1 - 2 - 3 371 64
JQ320285.1 - 2 - 3 320 64
JQ320286.1 - 2 - 3 320 64
JQ361659.1 - 2 - 3 320 64
KF026044.1 - 2 - 3 320 64
KJ776449.1 - 2 - 3 320 64
KJ776450.1 - 2 - 3 320 64
KU052691.1 - 2 - 3 320 64
KU052692.1 - 2 - 3 320 64
KU052693.1 - 2 - 3 320 64
KU052695.1 - 2 - 3 320 64
KU052696.1 - 2 - 3 320 64
KU052697.1 - 2 - 3 320 64
KU052698.1 - 2 - 3 320 64
KU052699.1 - 2 - 3 320 64
KU052700.1 - 2 - 3 320 64
KU052702.1 - 2 - 3 320 64
KU052703.1 - 2 - 3 320 64
LC002668.1 - 2 - 3 320 64
LC002669.1 - 2 - 3 320 64
LC002670.1 - 2 - 3 320 64
M92445.1 - 2 - 3 429 64
M93028.1 - 2 - 3 397 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0