This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03389170_m1
See other EIF4E2 GE Assays ›
Gene Symbol:
EIF4E2
Gene Name:
eukaryotic translation initiation factor 4E family member 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 133060054 - 133092624 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03389170_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF4E2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100191.2 NP_001093661.1 6 - 7 703 106
XM_013984576.2 XP_013840030.1 637 106
XM_021074503.1 XP_020930162.1 860 106
GenBank mRNA AK240297.1 - 6 - 7 702 106
AK343951.1 - 6 - 7 686 106
AK394397.1 - 6 - 7 720 106
DQ826509.1 - 6 - 7 704 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0