This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03385710_u1
See other RPL14 GE Assays ›
Gene Symbol:
RPL14 LOC100623913
Gene Name:
ribosomal protein L14
60S ribosomal protein L14
Gene Aliases:
-
RPL14
Chromosome Location:
Chr.13: 24691893 - 24698685 on Build Sscrofa11.1
Chromosome Location:
-
Species:
Pig
Species Specific ID (Flybase ID):
-
-
Assay ID Ss03385710_u1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001097478.1 NP_001090947.1 1 549 79
XM_021068170.1 XP_020923829.1 896 79
GenBank mRNA AK233622.1 - 6 555 79
AK347676.1 - 6 608 79
AK348520.1 - 6 609 79
AK394458.1 - 6 584 79
AK397607.1 - 6 555 79
AK400080.1 - 7 902 79
AY610461.1 - 1 534 79
AY610492.1 - 6 576 79
Gene Symbol: LOC100623913
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA DQ629165.1 - 1 549 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translation
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0