This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03385312_u1
See other IRF7 GE Assays ›
Gene Symbol:
IRF7
Gene Name:
interferon regulatory factor 7
Gene Aliases:
-
Chromosome Location:
Chr.2: 372235 - 375572 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03385312_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IRF7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001097428.1 NP_001090897.1 7 1447 129
XM_021076250.1 XP_020931909.1 1769 129
XM_021076252.1 XP_020931911.1 1645 129
GenBank mRNA AB287430.2 - 7 1447 129
AK235035.1 - 8 1656 129
EF195267.1 - 7 1236 129
EU294309.1 - 7 1236 129
KJ147515.1 - 7 1236 129

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Inventoried Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

winged helix/forkhead transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0