This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03380475_u1
See other AKR1E2 GE Assays ›
Gene Symbol:
AKR1E2
Gene Name:
aldo-keto reductase family 1 member E2
Gene Aliases:
AKR1CL2
Chromosome Location:
Chr.10: 65680314 - 65704701 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03380475_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AKR1E2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044568.3 NP_001038033.2 7 651 80
NR_145511.1 7 651 80
XM_013980351.2 XP_013835805.1 1065 80
XM_013980352.2 XP_013835806.1 933 80
XM_021064075.1 XP_020919734.1 1016 80
XM_021064076.1 XP_020919735.1 1112 80
GenBank mRNA AK394011.1 - 7 652 80
CB337983.1 - 7 651 80
DB808243.1 - 4 670 80
DQ474064.1 - 7 651 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
1,5-anhydro-D-fructose reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0