This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03378584_u1
See other PLA2G1B GE Assays ›
Gene Symbol:
PLA2G1B
Gene Name:
phospholipase A2 group IB
Gene Aliases:
PLA2, sPLA(2)-IB
Chromosome Location:
Chr.14: 40314520 - 40322315 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03378584_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLA2G1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004037.1 NP_001004037.1 3 - 4 352 76
GenBank mRNA M21055.1 - 3 - 4 352 76
Y00146.1 - 342 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
neutrophil mediated immunity
fatty acid biosynthetic process
positive regulation of cell proliferation
positive regulation of calcium ion transport into cytosol
lipid catabolic process
leukotriene biosynthetic process
neutrophil chemotaxis
interleukin-1 production
interleukin-8 production
cellular response to insulin stimulus
intracellular signal transduction
positive regulation of transcription from RNA polymerase II promoter
regulation of glucose import
phosphatidylcholine metabolic process
phosphatidylglycerol metabolic process
positive regulation of fibroblast proliferation
arachidonic acid secretion
positive regulation of immune response
positive regulation of NF-kappaB transcription factor activity
positive regulation of glomerular visceral epithelial cell apoptotic process
phospholipase A2 activity
receptor binding
calcium ion binding
bile acid binding
calcium-dependent phospholipase A2 activity
phospholipase A2 activity (consuming 1,2-dipalmitoylphosphatidylcholine)
phospholipase A2 activity consuming 1,2-dioleoylphosphatidylethanolamine)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0