This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03378336_u1
See other CDH5 GE Assays ›
Gene Symbol:
CDH5
Gene Name:
cadherin 5
Gene Aliases:
-
Chromosome Location:
Chr.6: 27108560 - 27142145 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03378336_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001649.2 NP_001001649.2 12 2156 105
GenBank mRNA EU714329.1 - 12 2097 105
Y13919.1 - 2156 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s)

blood vessel maturation
homophilic cell adhesion via plasma membrane adhesion molecules
regulation of complement-dependent cytotoxicity
regulation of establishment of cell polarity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0