This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03373371_g1
See other AIF1 GE Assays ›
Gene Symbol:
AIF1
Gene Name:
allograft inflammatory factor 1
Gene Aliases:
AIF-1
Chromosome Location:
Chr.7: 23711752 - 23731191 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03373371_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AIF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001129950.1 NP_001123422.1 4 - 5 273 68
XM_013977627.2 XP_013833081.1 400 68
XM_021098301.1 XP_020953960.1 671 68
GenBank mRNA AF299326.1 - 3 - 4 169 68
AK230615.1 - 4 - 5 274 68
AK230707.1 - 4 - 5 261 68
AK230962.1 - 4 - 5 274 68
AK231153.1 - 4 - 5 274 68
AK343645.1 - 4 - 5 274 68
AK343738.1 - 4 - 5 261 68
AK343889.1 - 4 - 5 278 68
AK343931.1 - 4 - 5 274 68
AK343963.1 - 4 - 5 274 68
AK343986.1 - 4 - 5 276 68
AK396271.1 - 4 - 5 420 68
EU282338.1 - 4 - 5 273 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

phagocytosis, engulfment
inflammatory response
Rac protein signal transduction
actin filament polymerization
positive regulation of T cell proliferation
positive regulation of smooth muscle cell proliferation
actin filament bundle assembly
cellular response to interferon-gamma
positive regulation of smooth muscle cell chemotaxis
positive regulation of monocyte chemotaxis
ruffle assembly
positive regulation of G1/S transition of mitotic cell cycle
positive regulation of T cell migration
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0