This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04173782_s1
See other SRO7 GE Assays ›
Gene Symbol:
SRO7
Gene Name:
Gene Aliases:
SNI1, SOP1, YPR032W
Chromosome Location:
Chr.XVI: 634123 - 637224 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04173782_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SRO7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001184129.1 NP_015357.1 1 3047 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
Golgi to plasma membrane transport
small GTPase mediated signal transduction
regulation of exocytosis
establishment of cell polarity
regulation of protein secretion
regulation of catalytic activity
SNARE binding
GTPase activator activity
syntaxin binding
small GTPase binding
myosin II binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0