This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04170000_s1
See other RAD1 GE Assays ›
Gene Symbol:
RAD1
Gene Name:
Gene Aliases:
LPB9, RAD12, YPL022W
Chromosome Location:
Chr.XVI: 506697 - 509999 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04170000_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001183836.1 NP_015303.1 1 3248 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

DNA metabolism protein endodeoxyribonuclease nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

meiotic mismatch repair
resolution of meiotic recombination intermediates
double-strand break repair via homologous recombination
removal of nonhomologous ends
double-strand break repair via single-strand annealing, removal of nonhomologous ends
DNA amplification
DNA repair
nucleotide-excision repair
nucleotide-excision repair, DNA incision, 5'-to lesion
mitotic recombination
cellular response to DNA damage stimulus
nucleotide-excision repair involved in interstrand cross-link repair
single-stranded DNA endodeoxyribonuclease activity
DNA binding
damaged DNA binding
single-stranded DNA binding
nuclease activity
endonuclease activity
endodeoxyribonuclease activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0