This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04164101_s1
See other MDH2 GE Assays ›
Gene Symbol:
MDH2
Gene Name:
Gene Aliases:
YOL126C
Chromosome Location:
Chr.XV: 81787 - 82920 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04164101_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MDH2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001183380.1 NP_014515.2 1 1050 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
tricarboxylic acid cycle
malate metabolic process
protein import into peroxisome matrix
carboxylic acid metabolic process
catalytic activity
oxidoreductase activity
malate dehydrogenase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
L-malate dehydrogenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0