This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04148598_s1
See other MCM5 GE Assays ›
Gene Symbol:
MCM5
Gene Name:
Gene Aliases:
BOB1, CDC46, YLR274W
Chromosome Location:
Chr.XII: 691555 - 693882 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04148598_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MCM5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001182161.1 NP_013376.1 1 2057 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via break-induced replication
DNA replication
pre-replicative complex assembly involved in nuclear cell cycle DNA replication
DNA replication initiation
transcription elongation from RNA polymerase II promoter
cell cycle
regulation of DNA-dependent DNA replication initiation
heterochromatin assembly
telomeric heterochromatin assembly
DNA duplex unwinding
nuclear DNA replication
nucleotide binding
DNA binding
DNA helicase activity
chromatin binding
DNA replication origin binding
single-stranded DNA binding
helicase activity
ATP binding
hydrolase activity
single-stranded DNA-dependent ATP-dependent DNA helicase activity
3'-5' DNA helicase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0