This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04130197_s1
See other MSR1 GE Assays ›
Gene Symbol:
MSR1
Gene Name:
Gene Aliases:
YHR091C
Chromosome Location:
Chr.VIII: 284839 - 286770 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04130197_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001179221.1 NP_011959.1 1 1847 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Gene Ontology Categories:

Function(s) Process(es)

translation
tRNA aminoacylation for protein translation
arginyl-tRNA aminoacylation
mitochondrial translation
mitochondrial arginyl-tRNA aminoacylation
nucleotide binding
aminoacyl-tRNA ligase activity
arginine-tRNA ligase activity
ATP binding
ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0