This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04123730_s1
See other SAE2 GE Assays ›
Gene Symbol:
SAE2
Gene Name:
Gene Aliases:
COM1, YGL175C
Chromosome Location:
Chr.VII: 173285 - 174322 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04123730_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SAE2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001181040.1 NP_011340.1 1 852 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

meiotic DNA double-strand break processing
telomere maintenance
DNA double-strand break processing
DNA catabolic process, endonucleolytic
DNA repair
cellular response to DNA damage stimulus
gene conversion at mating-type locus
DNA double-strand break processing involved in repair via synthesis-dependent strand annealing
telomeric 3' overhang formation
positive regulation of dephosphorylation
meiotic DNA double-strand break formation
meiotic cell cycle
positive regulation of exonuclease activity
single-stranded DNA endodeoxyribonuclease activity
DNA binding
double-stranded DNA binding
nuclease activity
endonuclease activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0