This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04112802_s1
See other DIN7 GE Assays ›
Gene Symbol:
DIN7
Gene Name:
Gene Aliases:
DIN3, YDR263C
Chromosome Location:
Chr.IV: 994242 - 995534 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04112802_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DIN7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001180571.3 NP_010549.3 1 947 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

exodeoxyribonuclease

Gene Ontology Categories:

Function(s) Process(es)

DNA double-strand break processing
DNA repair
mismatch repair
DNA recombination
cellular response to DNA damage stimulus
nucleic acid phosphodiester bond hydrolysis
DNA binding
catalytic activity
nuclease activity
endonuclease activity
hydrolase activity
hydrolase activity, acting on ester bonds
5'-flap endonuclease activity
5'-3' exodeoxyribonuclease activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0