This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Sc04103393_s1
See other DUT1 GE Assays ›
Gene Symbol:
DUT1
Gene Name:
Gene Aliases:
YBR252W
Chromosome Location:
Chr.II: 722611 - 723054 on Build R64
Species:
Saccharomyces cerevisiae
Species Specific ID (Flybase ID):
-
Assay ID Sc04103393_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DUT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001178600.3 NP_009811.3 1 155 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

dUMP biosynthetic process
nucleotide metabolic process
pyrimidine deoxyribonucleoside triphosphate catabolic process
dITP catabolic process
dUTP catabolic process
magnesium ion binding
dUTP diphosphatase activity
hydrolase activity
dITP diphosphatase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0