This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07363951_m1
See other HOXB8 GE Assays ›
Gene Symbol:
Hoxb8
Gene Name:
homeo box B8
Gene Aliases:
Hox2r1a
Chromosome Location:
Chr.10: 84131483 - 84133874 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07363951_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hoxb8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006247175.2 XP_006247237.1 1 - 2 437 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
grooming behavior
adult locomotory behavior
anterior/posterior pattern specification
sensory perception of pain
dorsal spinal cord development
negative regulation of myeloid cell differentiation
embryonic skeletal system morphogenesis
skeletal system morphogenesis
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0