This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07318644_g1
See other SVS3A GE Assays ›
Gene Symbol:
Svs3a LOC103694890
Gene Name:
seminal vesicle secretory protein 3A
seminal vesicle secretory protein 2-like
Gene Aliases:
Svs3
-
Chromosome Location:
Chr.3: 159082547 - 159084618 on Build Rnor_6.0
Chromosome Location:
Chr.Un|NW_007906879: 27812 - 30334 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn07318644_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Svs3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007605.1 NP_001007606.1 1120 94
GenBank mRNA D00920.1 - 1068 94
Gene Symbol: LOC103694890
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008773871.1 XP_008772093.1 1426 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Svs3a
Click here to find siRNAs for Gene LOC103694890

Gene Ontology Categories:

Function(s) Process(es)

fertilization
sexual reproduction
sperm capacitation
structural molecule activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0