This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn07314397_s1
See other MC3R GE Assays ›
Gene Symbol:
Mc3r
Gene Name:
melanocortin 3 receptor
Gene Aliases:
-
Chromosome Location:
Chr.3: 170252901 - 170253998 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn07314397_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mc3r
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025270.3 NP_001020441.3 1 1027 107
GenBank mRNA AY671938.1 - 1 1033 107
X70667.1 - 1 1225 107

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mc3r

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

regulation of heart rate
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
regulation of blood pressure
positive regulation of cAMP biosynthetic process
circadian regulation of gene expression
homoiothermy
locomotor rhythm
sodium ion homeostasis
regulation of feeding behavior
melanocortin receptor activity
melanocyte-stimulating hormone receptor activity
peptide hormone binding
neuropeptide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0